Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd3b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111336.1 | NP_001104806.1 | 1294 | |||
NM_008294.2 | NP_032320.1 | 1269 | ||||
GenBank mRNA | AK146749.1 | - | 4 | 1217 | ||
BC013449.1 | - | 4 | 1198 | 4221849 | ||
BC094029.1 | - | 4 | 1298 | |||
BC132402.1 | - | 4 | 1176 | 40130679 | ||
L16919.1 | - | 4 | 1210 |
Gene Symbol: Gm10681 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270429.1 | NP_001257358.1 | 4 | 1240 |