Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaja1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164671.1 | NP_001158143.1 | 2 | 305 | ||
NM_001164672.1 | NP_001158144.1 | 2 | 162 | |||
NM_008298.5 | NP_032324.1 | 2 | 256 | |||
GenBank mRNA | AF055664.1 | - | 2 | 171 | ||
AK049963.1 | - | 2 | 241 | |||
AK083046.1 | - | 2 | 192 | |||
AK145963.1 | - | 2 | 238 | |||
AK167139.1 | - | 2 | 192 | |||
AK167212.1 | - | 2 | 192 | |||
BC057876.1 | - | 2 | 167 | 5347227 | ||
BC060653.1 | - | 2 | 289 | |||
BC158024.1 | - | 2 | 122 | 9088153 | ||
BY245852.1 | - | 2 | 304 | |||
CK617936.1 | - | 2 | 162 | |||
CN669546.1 | - | 2 | 282 |