Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008314.2 | NP_032340.2 | 1, 2 | 1251 | ||
GenBank mRNA | AK137323.1 | - | 1, 2 | 126 | ||
AK141636.1 | - | 1, 2 | 1050 |
Set Membership: |
G-protein coupled receptor |