Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd79b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008339.2 | NP_032365.1 | 2, 3 | 171 | ||
GenBank mRNA | AK143573.1 | - | 2, 3 | 172 | ||
BC012226.1 | - | 2, 3 | 171 | 4164551 | ||
CT010358.1 | - | 2, 3 | 126 | |||
J03857.1 | - | 2, 3 | 310 |