Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008342.3 | NP_032368.2 | 2 | 633 | ||
GenBank mRNA | AK011784.1 | - | 3 | 504 | ||
AK011790.1 | - | 2 | 562 | |||
BC012724.1 | - | 2 | 621 | 2654907 | ||
BC054473.1 | - | 2 | 606 | 6488878 | ||
DV552718.1 | - | 1 | 161 | |||
X81580.1 | - | 2 | 604 |