Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hells | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008234.3 | NP_032260.2 | 4, 5 | 396 | ||
GenBank mRNA | AF155210.1 | - | 4, 5 | 396 | ||
AK013266.1 | - | 4, 5 | 435 | |||
AK088901.1 | - | 2, 3 | 1086 | |||
AK147126.1 | - | 4, 5 | 427 | |||
AK166636.1 | - | 4, 5 | 396 | |||
BC090671.1 | - | 4, 5 | 441 | |||
U25691.2 | - | 4, 5 | 429 |