Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hiat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008246.2 | NP_032272.2 | 3, 4 | 450 | ||
GenBank mRNA | AK004779.1 | - | 3, 4 | 389 | ||
AK135865.1 | - | 3, 4 | 271 | |||
AK158838.1 | - | 3, 4 | 451 | |||
AK169626.1 | - | 3, 4 | 390 | |||
BC118047.1 | - | 3, 4 | 270 | 40106779 | ||
D88315.1 | - | 3, 4 | 323 |