Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR0B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021969.2 | NP_068804.1 | 1 | 655 | ||
XM_011542297.2 | XP_011540599.1 | 1 | 655 | |||
GenBank mRNA | AK310048.1 | - | 1 | 656 | ||
BC030207.1 | - | 1 | 545 | 5222402 | ||
HQ692833.1 | - | 1 | 519 |