Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008306.4 | NP_032332.2 | 2 | 831 | ||
GenBank mRNA | AF049894.1 | - | 2 | 677 | ||
AF074926.1 | - | 2 | 677 | |||
AK035642.1 | - | 3 | 1240 | |||
AK147282.1 | - | 3 | 1224 | |||
AK156504.1 | - | 3 | 1185 | |||
BC066098.1 | - | 2 | 831 | 30533342 | ||
BC079561.1 | - | 2 | 831 | 30533342 |