Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Id3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008321.2 | NP_032347.1 | 3 | 772 | ||
GenBank mRNA | AK002820.1 | - | 3 | 771 | ||
AK014306.1 | - | 1 | 412 | |||
AK031499.1 | - | 1388 | ||||
AK192106.1 | - | 1 | 118 | |||
AK192467.1 | - | 1 | 118 | |||
AK199050.1 | - | 1 | 117 | |||
AK202308.1 | - | 1 | 117 | |||
M60523.1 | - | 3 | 780 |