Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hiat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008246.2 | NP_032272.2 | 9 | 1124 | ||
GenBank mRNA | AK004779.1 | - | 9 | 1063 | ||
AK041662.1 | - | 1 | 117 | |||
AK135865.1 | - | 9 | 945 | |||
AK158838.1 | - | 9 | 1125 | |||
AK169626.1 | - | 9 | 1064 | |||
BC118047.1 | - | 9 | 944 | 40106779 | ||
D88315.1 | - | 9 | 997 |