Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122950.2 | NP_001116422.1 | 2 | 686 | ||
NM_008263.3 | NP_032289.2 | 2 | 1090 | |||
GenBank mRNA | AK002670.1 | - | 2 | 238 | ||
AK188006.1 | - | 1 | 59 | |||
BC050839.1 | - | 2 | 1062 | 6511608 | ||
L08757.1 | - | 2 | 1090 |