Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008278.2 | NP_032304.2 | 2 | 196 | ||
GenBank mRNA | AK002472.1 | - | 2 | 164 | ||
AK146038.1 | - | 2 | 164 | |||
BB870447.1 | - | 2 | 197 | |||
BC021157.1 | - | 2 | 155 | 4208980 | ||
U44389.1 | - | 2 | 155 |