Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010973.2 | NP_001010973.1 | 2 | 1461 | ||
NM_008286.2 | NP_032312.2 | 2 | 1461 | |||
GenBank mRNA | AB444633.1 | - | 1 | 958 | ||
AB444634.1 | - | 1 | 958 | |||
AF513034.1 | - | 1 | 958 | |||
AF513035.1 | - | 1 | 958 | |||
AK020259.1 | - | 2 | 1462 | |||
AK163760.1 | - | 2 | 1458 | |||
BC101958.1 | - | 1 | 956 | |||
BC103552.1 | - | 1 | 957 | |||
BC103553.1 | - | 1 | 957 | |||
BC103554.1 | - | 1 | 957 |
Set Membership: |
G-protein coupled receptor |