Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaja1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164671.1 | NP_001158143.1 | 3, 4 | 538 | ||
NM_001164672.1 | NP_001158144.1 | 3, 4 | 395 | |||
NM_008298.5 | NP_032324.1 | 3, 4 | 489 | |||
GenBank mRNA | AF055664.1 | - | 3, 4 | 404 | ||
AK049963.1 | - | 3, 4 | 474 | |||
AK083046.1 | - | 3, 4 | 425 | |||
AK145963.1 | - | 3, 4 | 471 | |||
AK167139.1 | - | 3, 4 | 425 | |||
AK167212.1 | - | 3, 4 | 425 | |||
BC057876.1 | - | 3, 4 | 400 | 5347227 | ||
BC060653.1 | - | 3, 4 | 522 | |||
BC158024.1 | - | 3, 4 | 355 | 9088153 | ||
CK617936.1 | - | 3, 4 | 395 | |||
CN669546.1 | - | 3, 4 | 515 |