Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293557.1 | NP_001280486.1 | 2 | 685 | ||
NM_022162.2 | NP_071445.1 | 3 | 725 | |||
XM_005256084.3 | XP_005256141.1 | 3 | 629 | |||
XM_006721242.3 | XP_006721305.1 | 3 | 629 | |||
XM_006721243.3 | XP_006721306.1 | 3 | 629 | |||
XM_011523260.2 | XP_011521562.1 | 3 | 629 | |||
XM_011523261.1 | XP_011521563.1 | 3 | 629 | |||
XM_017023536.1 | XP_016879025.1 | 2 | 156 | |||
XM_017023538.1 | XP_016879027.1 | 2 | 151 | |||
GenBank mRNA | AF178930.1 | - | 3 | 725 | ||
AY187243.1 | - | 3 | 725 | |||
AY187244.1 | - | 3 | 1447 | |||
DQ868973.1 | - | 3 | 607 | |||
HQ204571.1 | - | 3 | 607 |