Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008314.2 | NP_032340.2 | 2 | 1560 | ||
GenBank mRNA | AK137323.1 | - | 2 | 435 | ||
AK141636.1 | - | 2 | 1359 | |||
BC115911.1 | - | 2 | 1252 | 40104765 | ||
BC116654.1 | - | 2 | 1252 | 40104769 |
Set Membership: |
G-protein coupled receptor |