Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131055.1 | NP_001124527.1 | 2 | 1183 | ||
NM_022304.2 | NP_071640.1 | 1 | 2312 | |||
XM_005265904.3 | XP_005265961.1 | 2 | 1457 | |||
XM_006714864.3 | XP_006714927.1 | 4 | 1686 | |||
XM_006714865.3 | XP_006714928.1 | 2 | 1458 | |||
XM_011534548.2 | XP_011532850.1 | 3 | 1594 | |||
XM_011534549.2 | XP_011532851.1 | 4 | 1636 | |||
XM_017009426.1 | XP_016864915.1 | 2 | 1595 | |||
GenBank mRNA | AB451483.1 | - | 1 | 539 | ||
AY136744.1 | - | 1 | 539 | |||
BC054510.2 | - | 2 | 1197 | 6146988 |
Set Membership: |
G-protein coupled receptor |