Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008331.3 | NP_032357.2 | 2 | 553 | ||
GenBank mRNA | AK131669.1 | - | 2 | 551 | ||
AK144230.1 | - | 2 | 558 | |||
AK150352.1 | - | 2 | 560 | |||
AK150424.1 | - | 2 | 554 | |||
AK150609.1 | - | 2 | 563 | |||
AK151125.1 | - | 2 | 563 | |||
AK151158.1 | - | 2 | 554 | |||
AK151278.1 | - | 2 | 563 | |||
AK151298.1 | - | 2 | 557 | |||
AK151983.1 | - | 2 | 554 | |||
AK152112.1 | - | 2 | 556 | |||
AK152753.1 | - | 2 | 554 | |||
AK152853.1 | - | 1 | 228 | |||
AK153184.1 | - | 2 | 556 | |||
AK153500.1 | - | 2 | 554 | |||
AK153532.1 | - | 2 | 563 | |||
AK157848.1 | - | 2 | 554 | |||
BC003768.1 | - | 2 | 537 | 3496673 | ||
CT010226.1 | - | 1 | 459 | |||
U43084.1 | - | 2 | 528 |