Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il10rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008349.5 | NP_032375.2 | 2 | 175 | ||
GenBank mRNA | AF440787.1 | - | 2 | 102 | ||
AK040938.1 | - | 2 | 91 | |||
AK134042.1 | - | 2 | 168 | |||
AK160991.1 | - | 2 | 1130 | |||
BC145791.1 | - | 2 | 175 | 40131094 | ||
U53696.1 | - | 2 | 88 |