Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008361.3 | NP_032387.1 | 3, 4 | 172 | ||
GenBank mRNA | AK156396.1 | - | 2, 3 | 724 | ||
AK157245.1 | - | 2, 3 | 724 | |||
AK168047.1 | - | 3, 4 | 192 | |||
AK224980.1 | - | 2, 3 | 724 | |||
AK225002.1 | - | 3, 4 | 191 | |||
BC011437.1 | - | 3, 4 | 172 | 3989461 | ||
M15131.1 | - | 3, 4 | 181 |