Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008396.2 | NP_032422.2 | 2 | 213 | ||
GenBank mRNA | AK036572.1 | - | 2 | 273 | ||
AK051050.1 | - | 2 | 224 | |||
AK156604.1 | - | 2 | 255 | |||
BC065139.1 | - | 2 | 213 | 30619401 | ||
Z29987.1 | - | 2 | 217 |