Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277970.1 | NP_001264899.1 | 1 | 298 | ||
NM_008397.4 | NP_032423.2 | 1 | 298 | |||
GenBank mRNA | AK018033.1 | - | 1 | 308 | ||
AK034186.1 | - | 1 | 155 | |||
BC049132.1 | - | 2 | 223 | |||
BC058095.1 | - | 2 | 223 | 6826609 | ||
X69902.1 | - | 1 | 279 |