Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008404.4 | NP_032430.2 | 3 | 192 | ||
GenBank mRNA | AK088435.1 | - | 3 | 181 | ||
AK136502.1 | - | 3 | 193 | |||
AK161236.1 | - | 3 | 161 | |||
AK169587.1 | - | 3 | 167 | |||
BC145644.1 | - | 2 | 114 | 40130947 | ||
M31039.1 | - | 2 | 80 | |||
X14951.1 | - | 3 | 158 |