Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008420.4 | NP_032446.2 | 1 | 440 | ||
GenBank mRNA | BC031776.1 | - | 1 | 396 | 4507847 | |
BC051422.1 | - | 1 | 397 | 6493266 | ||
BC061501.1 | - | 1 | 421 | 4506061 | ||
DQ646376.1 | - | 694 | ||||
DQ646377.1 | - | 1 | 440 | |||
EF437378.1 | - | 1 | 230 | |||
M64228.1 | - | 1 | 245 |