Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008439.3 | NP_032465.2 | 4 | 398 | ||
GenBank mRNA | BC013464.1 | - | 4 | 398 | 4235560 | |
Y09335.1 | - | 4 | 377 |