Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008360.1 | NP_032386.1 | 5 | 475 | ||
GenBank mRNA | AY157834.1 | - | 5 | 468 | ||
AY157835.1 | - | 5 | 468 | |||
AY362457.1 | - | 4 | 320 | |||
BC024384.1 | - | 5 | 469 | 4988988 | ||
D49949.1 | - | 5 | 475 | |||
DQ316604.1 | - | 4 | 311 | |||
U66244.1 | - | 4 | 311 |