Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008371.4 | NP_032397.1 | 3 | 412 | ||
GenBank mRNA | AK040399.1 | - | 3 | 196 | ||
AK041307.1 | - | 3 | 413 | |||
AK041403.1 | - | 114 | ||||
AK042009.1 | - | 5 | 1197 | |||
AK052452.1 | - | 4 | 1100 | |||
AK139858.1 | - | 4 | 773 | |||
AK145184.1 | - | 3 | 703 | |||
BC110553.2 | - | 3 | 321 | 40047584 | ||
X07962.1 | - | 3 | 712 |