Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008391.4 | NP_032417.3 | 2, 3 | 293 | ||
GenBank mRNA | AK036652.1 | - | 2, 3 | 261 | ||
AK087926.1 | - | 2, 3 | 279 | |||
AK132099.1 | - | 2, 3 | 306 | |||
AK155025.1 | - | 2, 3 | 278 | |||
BC006577.1 | - | 2, 3 | 286 | 3586586 | ||
BC110666.1 | - | 2, 3 | 265 | 4918447 | ||
J03168.1 | - | 2, 3 | 248 |