Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008445.2 | NP_032471.2 | 1 | 1361 | ||
GenBank mRNA | AF013116.1 | - | 1 | 937 | ||
AK133649.1 | - | 1 | 1343 | |||
AK147572.1 | - | 1 | 1163 | |||
AK220283.1 | - | 2 | 1086 | |||
BC052732.1 | - | 1 | 518 | |||
BC053053.1 | - | 1 | 1303 | |||
BC062648.1 | - | 1 | 1036 | |||
BC127063.1 | - | 1 | 573 | 40109965 |