Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008448.3 | NP_032474.2 | 4 | 735 | ||
GenBank mRNA | AK133535.1 | - | 4 | 732 | ||
AK136307.1 | - | 4 | 732 | |||
AK177424.1 | - | 4 | 233 | |||
AK185672.1 | - | 3 | 207 | |||
BC013248.1 | - | 4 | 691 | |||
BC013817.1 | - | 4 | 701 | |||
BC025864.1 | - | 4 | 704 | |||
BC040800.1 | - | 4 | 725 | |||
BC069920.1 | - | 4 | 711 | |||
BC090841.1 | - | 4 | 711 | 30543821 | ||
L27153.1 | - | 4 | 329 | |||
U86090.1 | - | 4 | 562 |