Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008451.2 | NP_032477.2 | 2 | 384 | ||
GenBank mRNA | AF055666.1 | - | 2 | 220 | ||
AK134796.1 | - | 1 | 59 | |||
AK141439.1 | - | 2 | 314 | |||
BC014845.1 | - | 2 | 228 | 2599082 | ||
BQ887879.1 | - | 2 | 252 | |||
CB524468.1 | - | 2 | 536 |