Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il10ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008348.2 | NP_032374.1 | 7 | 2245 | ||
GenBank mRNA | AK137663.1 | - | 7 | 2246 | ||
AK149680.1 | - | 7 | 2240 | |||
AK149899.1 | - | 6 | 2128 | |||
AK150022.1 | - | 7 | 2242 | |||
AK150524.1 | - | 7 | 2250 | |||
AK153120.1 | - | 7 | 2249 | |||
AK159566.1 | - | 7 | 2242 | |||
L12120.1 | - | 7 | 2250 |