Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il13ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089687.2 | - | 9 | 1398 | ||
BC003723.1 | - | 6 | 574 | 3486223 | ||
EF219410.1 | - | 4 | 413 | |||
U65747.1 | - | 6 | 662 |