Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il15ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271497.1 | NP_001258426.1 | 4, 5 | 656 | ||
NM_001271498.1 | NP_001258427.1 | 3, 4 | 679 | |||
NM_001271499.1 | NP_001258428.1 | 4, 5 | 620 | |||
NM_008358.2 | NP_032384.1 | 4, 5 | 719 | |||
NM_133836.2 | NP_598597.1 | 4, 5 | 772 | |||
GenBank mRNA | AK040940.1 | - | 3, 4 | 531 | ||
AK080000.1 | - | 3, 4 | 531 | |||
AY219715.1 | - | 4, 5 | 521 | |||
AY221616.1 | - | 4, 5 | 397 | |||
AY221619.1 | - | 3, 4 | 418 | |||
BC022705.1 | - | 4, 5 | 529 | |||
BC095982.1 | - | 1, 2 | 447 | |||
BC132231.1 | - | 3, 4 | 319 | |||
BC132233.1 | - | 3, 4 | 319 | |||
BC145348.1 | - | 3, 4 | 319 |