Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIVEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127714.2 | NP_001121186.1 | 3 | 1098 | ||
NM_024503.4 | NP_078779.2 | 4 | 1178 | |||
XM_011541884.2 | XP_011540186.1 | 5 | 1110 | |||
XM_011541885.2 | XP_011540187.1 | 4 | 1390 | |||
XM_011541886.2 | XP_011540188.1 | 4 | 1390 | |||
XM_011541887.2 | XP_011540189.1 | 4 | 1390 | |||
XM_017001992.1 | XP_016857481.1 | 5 | 2357 | |||
XM_017001993.1 | XP_016857482.1 | 3 | 1310 | |||
XM_017001994.1 | XP_016857483.1 | 4 | 1390 | |||
GenBank mRNA | AB046775.3 | - | 3 | 1098 | ||
AF278765.1 | - | 4 | 971 | |||
BC152563.1 | - | 3 | 1098 | 8860523 |