Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il2rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008368.4 | NP_032394.1 | 6 | 628 | ||
GenBank mRNA | AK017288.1 | - | 6 | 628 | ||
AK035289.1 | - | 6 | 581 | |||
AK157572.1 | - | 6 | 629 | |||
AK169481.1 | - | 6 | 609 | |||
BC026869.1 | - | 6 | 601 | 4015306 | ||
M28052.1 | - | 6 | 585 |