Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il7r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008372.4 | NP_032398.3 | 8 | 2255 | ||
GenBank mRNA | AK154215.1 | - | 7 | 2069 | ||
AK154516.1 | - | 8 | 2216 | |||
BC089571.1 | - | 8 | 2190 | 6758487 | ||
M29697.1 | - | 8 | 2161 |