Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008373.1 | NP_032399.1 | 5 | 378 | ||
GenBank mRNA | X14045.1 | - | 5 | 378 |