Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005109.2 | NP_005100.1 | 18 | 4372 | ||
XM_017007601.1 | XP_016863090.1 | 17 | 4179 | |||
GenBank mRNA | AB017642.1 | - | 18 | 4372 | ||
AB029024.1 | - | 18 | 4125 | |||
BC008726.2 | - | 18 | 4355 | 3163379 | ||
BI090706.1 | - | 1 | 256 |