Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008391.4 | NP_032417.3 | 9 | 2149 | ||
GenBank mRNA | AK036652.1 | - | 9 | 2124 | ||
AK087926.1 | - | 8 | 2017 | |||
AK132099.1 | - | 9 | 2163 | |||
AK155025.1 | - | 9 | 2135 | |||
BC006577.1 | - | 9 | 2140 | 3586586 | ||
BC110666.1 | - | 9 | 2123 | 4918447 | ||
J03168.1 | - | 9 | 2102 |