Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008410.2 | NP_032436.1 | 6 | 1186 | ||
GenBank mRNA | AB030203.1 | - | 6 | 1207 | ||
AK002796.1 | - | 1 | 341 | |||
AK005125.1 | - | 6 | 1177 | |||
AK008861.1 | - | 1 | 341 | |||
AK076139.1 | - | 6 | 1195 | |||
AK088270.1 | - | 7 | 1176 | |||
AK150378.1 | - | 6 | 1206 | |||
AK150396.1 | - | 6 | 1205 | |||
AK151714.1 | - | 6 | 1206 | |||
AK151889.1 | - | 6 | 1206 | |||
AK152068.1 | - | 6 | 1194 | |||
AK152516.1 | - | 6 | 1206 | |||
AK152650.1 | - | 6 | 1206 | |||
AK152983.1 | - | 6 | 1198 | |||
AK153103.1 | - | 6 | 1149 | |||
AK159307.1 | - | 6 | 1150 | |||
AK159340.1 | - | 6 | 1174 | |||
AK159378.1 | - | 6 | 1205 | |||
AK159426.1 | - | 6 | 1174 | |||
AK159619.1 | - | 6 | 1198 | |||
AK159792.1 | - | 6 | 1174 | |||
AK159962.1 | - | 6 | 1177 | |||
AK160101.1 | - | 6 | 1174 | |||
AK160725.1 | - | 6 | 1205 | |||
AK205546.1 | - | 1 | 225 | |||
AK205709.1 | - | 1 | 225 | |||
AK215843.1 | - | 1 | 227 | |||
AK217202.1 | - | 1 | 224 | |||
AK218926.1 | - | 1 | 225 | |||
BC004731.1 | - | 6 | 1157 | 3583154 | ||
U76253.1 | - | 6 | 1186 |