Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008417.5 | NP_032443.3 | 3 | 3233 | ||
GenBank mRNA | AK044342.1 | - | 3 | 3235 |