Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008424.3 | NP_032450.1 | 2 | 2125 | ||
GenBank mRNA | AK028907.1 | - | 2 | 2126 |