Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008425.4 | NP_032451.1 | 2 | 1843 | ||
GenBank mRNA | AK044915.1 | - | 2 | 1844 | ||
BC137841.1 | - | 1 | 1613 | 8860860 | ||
BC137842.1 | - | 1 | 1613 | 8860861 | ||
X73052.1 | - | 2 | 1774 |