Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008439.3 | NP_032465.2 | 5, 6 | 590 | ||
GenBank mRNA | AK002716.1 | - | 1, 2 | 30 | ||
AK008326.1 | - | 1, 2 | 133 | |||
AK008503.1 | - | 1, 2 | 133 | |||
BC013464.1 | - | 5, 6 | 590 | 4235560 | ||
Y09335.1 | - | 5, 6 | 569 |