Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008444.4 | NP_032470.3 | 2 | 692 | ||
GenBank mRNA | AK081541.1 | - | 2 | 650 | ||
AK083700.1 | - | 2 | 680 | |||
AK122258.1 | - | 2 | 653 | |||
AK147467.1 | - | 2 | 686 | |||
AK148200.1 | - | 2 | 673 | |||
BC065132.1 | - | 2 | 659 | |||
BC139388.1 | - | 1 | 611 | 8862410 | ||
BC139389.1 | - | 1 | 611 | 8862411 | ||
CJ089315.1 | - | 2 | 694 | |||
D26077.1 | - | 1 | 605 |