Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130150.1 | NP_001123622.1 | 3 | 244 | ||
NM_001130151.1 | NP_001123623.1 | 3 | 244 | |||
NM_001130152.1 | NP_001123624.1 | 3 | 253 | |||
NM_001130153.1 | NP_001123625.1 | 3 | 244 | |||
NM_008488.2 | NP_032514.1 | 3 | 244 | |||
GenBank mRNA | AK012156.1 | - | 3 | 194 | ||
AK043027.1 | - | 3 | 208 | |||
AK051518.1 | - | 3 | 194 | |||
AK131181.1 | - | 3 | 141 | |||
AK157056.1 | - | 3 | 180 | |||
AK162482.1 | - | 3 | 157 | |||
AK172354.1 | - | 3 | 180 | |||
AK211978.1 | - | 1 | 12 | |||
BB840025.1 | - | 3 | 244 | |||
BC012488.1 | - | 3 | 204 | 3154558 | ||
BY323306.1 | - | 3 | 254 | |||
CV783191.1 | - | 3 | 176 | |||
U58203.1 | - | 3 | 174 |