Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028796.1 | - | 3 | 948 | ||
AK156499.1 | - | 2 | 827 | |||
AK157297.1 | - | 2 | 817 | |||
AK165124.1 | - | 1 | 56 | |||
AK171128.1 | - | 3 | 891 | |||
AK172551.1 | - | 2 | 817 | |||
AK172573.1 | - | 2 | 821 | |||
BC006759.1 | - | 2 | 801 | 3709861 | ||
U89992.2 | - | 712 |