Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008533.2 | NP_032559.2 | 2, 3 | 407 | ||
GenBank mRNA | AK089255.1 | - | 2, 3 | 398 | ||
AK156548.1 | - | 2, 3 | 398 | |||
D37797.1 | - | 2, 3 | 377 |